Learn More
DuPont™ Tyvek 500 Collared Laboratory Coat
Product Code. 12322249 Shop All DuPont Products
Change view
Click to view available options
Size: Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12322249 Size: Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12322249 Supplier DuPont™ Supplier No. D13496004
  • ONLINE EXCLUSIVE PRICE
    85.93 EUR
    68.27 EUR / Pack of 10
    Save 17.66 EUR
    20% Off
Estimated Shipment: 01-12-2026
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Zipper and Pockets are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
  • Silicon non-added
  • With collar, zipper closure and 2 pockets
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY
Specifications
Material HDPE
Color White
Closure Type Zipper
Resistance Chemicals
No. of Pockets 2
Seam Type Stitched (internal)
Certifications/Compliance Antistatic treatment (EN 1149-5)
For Use With (Application) Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others