Learn More
DuPont™ Tyvek™ 500 Xpert (Eco Pack)
Product Code. 17563266 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 100
Pack of 25
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17563266 3X-Large Pack of 25
17513266 Small Pack of 25
17301609 Small Pack of 100
17523266 Medium Pack of 25
17311609 Medium Pack of 100
17533266 Large Pack of 25
17321609 Large Pack of 100
17543266 X-Large Pack of 25
17331609 X-Large Pack of 100
17553266 2X-Large Pack of 25
17341609 2X-Large Pack of 100
17351609 3X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
Artikelnummer. 17563266 Lieferant DuPont™ Lieferanten-Nr. D15359284
  • ONLINE EXCLUSIVE PRICE
    278.99 EUR
    201.40 EUR / Pack of 25
    Save 77.59 EUR
    27% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
Hooded coverall with elasticated waist and ergonomic and protective design.
Hooded coverall is robust yet lightweight at less than 180 grams per garment. Its 3-piece hood provides an optimal fit to the head and face when turning. Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek™ is permeable to both air and water vapor, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibers (down to 1 micron in size), is ultra-low-linting and antistatically treated.
Applications for Tyvek™ 500 Xpert garments include those found in pharmaceutical handling, chemical processing, the oil and gas industry, general maintenance and operations, automatic spray painting and more.
  • No silicone added
  • Elasticated face, wrists and ankles
  • Glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper pull
  • Tyvek™ auto-lock zipper and zipper flap for increased protection
  • Chemical protective clothing Category III, Type 5-B and 6-B
  • Protection against radioactive contamination
  • Antistatic treatment on both sides
  • Stitched external seams
  • Very low inward leakage due to optimized design
With Eco Pack, there are 25 garments in a value pack together with one IFU, instead of an individual packaging together with one IFU for each garment unit. The product and its performance remain exactly the same but the changes in packaging, reduce the use of paper and plastic and thus help to better protect the planet.
TRUSTED_SUSTAINABILITY
Spezifikation
Size 3X-Large
Gender Unisex
Material Polyethylene
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Seam Type Stitched
Sterility Non-sterile
Protection Class Category III
Resistance Chemicals
Certifications/Compliance (Europe) Regulation (EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
Latex Free No
Weight (English) <6.3 oz.
Weight (Metric) <180 g
Mehr anzeigen Weniger anzeigen