missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin AG 528, MedChemExpress
Description
Tyrphostin AG 528 is an inhibitor of EGFR and ErbB2 with IC50s of 4.9 and 2.1 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Tyrphostin AG 528 |
| CAS | 133550-49-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H14N2O3 |
| Quantity | 100 mg |
| Synonym | Tyrphostin B66 AG 528 |
| Solubility Information | DMSO : 25 mg/mL (81.61 mM; Need ultrasonic) |
| SMILES | O=C(/C(C#N)=C/C1=CC=C(O)C(O)=C1)N2CCC3=CC=CC=C32 |
| Molecular Weight (g/mol) | 306.32 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?