missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Tyrosine Kinase Adaptors Phospho-Specific Array
Click to view available options
Quantity:
1 Kit (2 Samples)
Unit Size:
1 set
Description
- This product must be accompanied with Antibody Array Detection Kit
- The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
- Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Specifications
Specifications
| Content And Storage | 4°C |
| Detection Method | Fluorometric |
| Quantity | 1 Kit (2 Samples) |
| For Use With (Application) | Profiling |
| Product Type | Array/Library |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction