Description
- Double cuffs for increased protection and tighter connection with gloves
- Elasticated wrists
- Knitted cuff (inside) for better comfort
- Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
- Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
- Disposable
Specifications
Specifications
| Size | 2X-Large |
| Color | Yellow |
| Disposable | Single-use |
| Certifications/Compliance | (EU) 2016/425, EN 14126, EN 1149-1 |