Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Shop All Thermo Scientific Products
Click to view available options
Capacity:
1.5/2 mL Conical Microtube
100 mL Round-bottom Open-top Tube
14 mL Conical Urine Tube
15 mL Conical Tube
25 mL Universal Conical/Skirted Tube
250 mL Bottle
250 mL Corning Conical Bottle
5 mL RIA/Round-bottom Tube (w/o cap)
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid
50 mL Conical Tube
50 mL Round-bottom Tube
84 x 15 mL Blood Collection Tube
Unit Size:
Pack of 4
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
Specifications
| Type | Round Bucket Adapter |
| Tube Type | Microtube, Conical |
| Capacity | 1.5/2 mL Conical Microtube |
| For Use With (Equipment) | Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube |
| Tube Size | 1.5/2 mL |