Learn More
Köttermann Two Door Tall Storage Cabinet
Product Code. 17362089
Change view
Click to view available options
Door Type:
Hinged
Sliding
No. of Shelves:
4
None
Dimensions (D x W x H):
350 x 900 x 1920 mm
366 x 900 x 1920 mm
500 x 900 x 1920 mm
516 x 900 x 1920 mm
350 x 1200 x 1920 mm
500 x 1200 x 1920 mm
770 x 1100 x 2250 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Type No. of Shelves Dimensions (D x W x H) unitSize
17362089 Sliding 4 350 x 900 x 1920 mm Each
17342079 Sliding 4 350 x 1200 x 1920 mm Each
17382089 Hinged 4 366 x 900 x 1920 mm Each
17392089 Sliding 4 500 x 900 x 1920 mm Each
17362079 Sliding 4 500 x 1200 x 1920 mm Each
17312099 Hinged 4 516 x 900 x 1920 mm Each
17322099 Hinged None 770 x 1100 x 2250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17362089 Supplier Köttermann Supplier No. 30100066
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
This Two Door Tall Storage Cabinet features a two hinged or sliding doors to protect stored equipment and utensils.
  • Tall, slender design expands storage with a small footprint
  • Two doors protect stored items
  • Sliding and hinged door options available
Specifications
Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 350 x 900 x 1920 mm
Door Type Sliding
Finish Epoxy Coated
For Use With (Application) Storage
No. of Doors 2
No. of Drawers None
No. of Shelves 4
Warranty None
Weight (Metric) 83.4 kg
With Casters No
Material Steel