Learn More
Brannan™ Twin Reading Digital Thermometer
Product Code. 17771318
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +50°C, -50°C to +70°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
17771318 -20°C to +50°C, -50°C to +70°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17771318 Supplier Brannan™ Supplier No. 22/420/3
  • ONLINE EXCLUSIVE PRICE
    12.35 EUR
    10.50 EUR / Pack of 1
    Save 1.85 EUR
    14% Off
Estimated Shipment: 08-01-2027
Brannan Digital Thermometer with max/min feature and high/low temperature alarm setting.
  • Twin reading thermometer ideally suited to read temperature in a fridge or freezer or outdoor temperature and room temperature simultaneously
  • Max min recording facility
  • Celsius and fahrenheit switchable
  • 2 m cable to external sensor
  • Magnetic fixture on back of meter
  • -20°C to +50°C/0°F to 120°F
  • 1x AAA battery supplied
Specifications
Display Dimensions 21 x 47 mm
Disposable Reusable
Emissivity No
For Use With (Application) Read Temp in a Fridge or Freezer
Height (English) 2.48 in.
Height (Metric) 6.3 cm
Includes 2 m Cable to External Sensor
Length (Metric) 6.3 cm
Material Plastic
Measurement Time 10 sec.
Model 22/420/3
Probe Type 2 m Cable to External Sensor
Resolution 0.1°C
Weight (English) 0.13 lb.
Weight (Metric) 62 g
Width (English) 0.59 in.
Data Storage Data logging
Max. Temperature (English) 120°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 0°F
Min. Temperature (Metric) -20°C
Temperature Range (English) Internal Temp -4°F to +122°F, External Temp -58°F to +158°F
Temperature Range (Metric) -20°C to +50°C, -50°C to +70°C
For Use With (Equipment) Fridge or Freezer
Battery Life 2 to 3 yr.
Battery Type AAA
Accuracy ±1.0% between -20°C and +70°C
Display LCD
Width (Metric) 1.5 cm