Learn More
TWEEN™ 85, Sigma-Aldrich™
Specifications
Specifications
| CAS | 9005-70-3 |
| Density | 1.03 g/cm3 at 20°C |
| Concentration | 100% |
| Quantity | 500 mL |
| MDL Number | MFCD01779641 |
| Chemical Name or Material | TWEEN™ 85 |