missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEEN™ 80, Sigma-Aldrich™
Specifications
Specifications
| Boiling Point | >100°C/1013 hPa |
| Density | 1.07 g/cm3 at 25°C |
| Concentration | 100% |
| Quantity | 2.5 L |
| MDL Number | MFCD00082107 |
| Synonym | POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80 |
| Grade | Ungraded |
| Chemical Name or Material | Tween™ 80 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?