missing translation for 'onlineSavingsMsg'
Learn More

TWEEN™ 40, Sigma-Aldrich™

Product Code. 15891701
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891701 500 mL 500mL
15801711 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15891701 Supplier Sigma-Aldrich Supplier No. 8221850500
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-66-7

Specifications

Boiling Point >100°C
Density 1.09 g/cm3 at 20°C
Concentration 100%
Quantity 500 mL
MDL Number MFCD00165345
Grade Ungraded
Chemical Name or Material Tween™ 40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.