missing translation for 'onlineSavingsMsg'
Learn More

Tungstophosphoric acid hydrate, Supelco™

Product Code. 32238094
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238094 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238094 Supplier Supelco Supplier No. 1005829025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Cryst. EMPLURA™

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungstophosphoric acid hydrate
CAS 12501-23-4
Melting Point 107°C
Molecular Formula H3[P(W3O10)4] · x H2O
Quantity 25 kg
Synonym Tungstophosphoric acid hydrate, Phosphotungstic acid, Tungstophosphoric acid hydrate
Molecular Weight (g/mol) 2880.05 (anhydrous basis)
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.