Learn More
Tungstophosphoric Acid Hydrate, Supelco™
Product Code. 15896860
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896860 250 g 250g
15886860 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896860 Supplier Supelco Supplier No. 1005830250
 Restricted Item
Estimated Shipment: 23-11-2026
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12501-23-4
Specifications
Melting Point 107°C
Quantity 250 g
Molecular Formula H3[P(W3O10)4] · X H2O
MDL Number MFCD00149915
Synonym Tungstophosphoric acid hydrate, Phosphotungstic acid, Tungstophosphoric acid hydrate
Molecular Weight (g/mol) 2880.05 (anhydrous basis)
Grade Ungraded
Chemical Name or Material Tungstophosphoric Acid Hydrate