Learn More
SPEX™ SamplePrep Tungsten Carbide Grinding Ball Set of 4
Product Code. 15330532 Shop All SPEX Certiprep Products
Change view
Click to view available options
Capacity (Metric):
5 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15330532 5 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15330532 Supplier SPEX™ SamplePrep Supplier No. 5004A
SPEX SamplePrep™ Replacement Tungsten carbide ball for 5004 Tungsten Carbide Grinding Vial Set.
  • Replacement tungsten carbide grinding ball for 5004 Tungsten Carbide Lined Grinding Vial Set
  • Sold as a bag of 4 grinding balls
Specifications
Product Type Grinding Vial Accessory
Capacity (Metric) 5 mL
For Use With (Equipment) 5004
Height (Metric) 7.9 mm
Includes 4 grinding balls
Length (Metric) 7.9 mm
Material Tungsten Carbide
Width (Metric) 7.9 mm