Learn More
Palintest™ Tubetest COD Reagent Tube with Mercury For High Chloride Sample
Product Code. 11938937
Change view
Click to view available options
Range:
0 to 150mg/L
0 to 20000mg/L
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11938937 0 to 20000mg/L Pack of 24
11918937 0 to 150mg/L Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11938937 Supplier Palintest™ Supplier No. PL467
  • ONLINE EXCLUSIVE PRICE
    112.89 EUR
    71.30 EUR / Pack of 24
    Save 41.59 EUR
    36% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Suitable for working with samples with very high chloride levels, this high chloride formulation (2M), is ideal for working in laboratory and other types of scientific environments.
  • The mercury free formulation is recommended for general use 
  • The mercuric sulfate formulation (M) is provided for use in standard procedures which require the suppression of interference due to moderate levels of chloride
  • The high chloride formulation (2M) provides much higher levels of chloride suppression and is suitable for working with samples with very high chloride levels
  • The new M/C formulation with mercuric sulfate is specifically for use in non-Palintest™ instruments
Specifications
For Use With (Application) High Chloride Samples Such as Chemical Oxygen Demand (COD)
For Use With (Equipment) Photometer or UV-Vis Spectrophotometer
Includes Chemical Oxygen Demand (COD)
No. of Tests 25
Range 0 to 20000mg/L
Type COD Reagent Tube