Learn More
Tubeimoside A, TRC
Specifications
Specifications
| Chemical Name or Material | Tubeimoside A |
| CAS | 102040-03-9 |
| Molecular Formula | C63 H98 O29 |
| Quantity | 100 mg |
| Synonym | Olean-12-en-28-oic acid','3-[[2-O-[4-O-[(3S)-4-carboxy-3-hydroxy-3-methyl-1-oxobutyl]-α-L-arabinopyranosyl]-β-D-glucopyranosyl]oxy]-2,23-dihydroxy-','28-(O-β-D-xylopyranosyl-(1→3)-O-6-deoxy-α-L-mannopyranosyl-(1→2)-α-L-arabinopyranosyl) ester','intramol. 4''(3)→4''(28)-ester','(2β,3β,4α)-, Olean-12-en-28-oic acid','3-[[2-O-[4-O-(4-carboxy-3-hydroxy-3-methyl-1-oxobutyl)-α-L-arabinopyranosyl]-β-D-glucopyranosyl]oxy]-2,23-dihydroxy-','28-(O-β-D-xylopyranosyl-(1→3)-O-6-deoxy-α-L-mannopyranosyl-(1→2)-α-L-arabinopyranosyl) ester','intramol. ester','[2β,3β(S),4α]-, Lobatoside H, Tubeimoside 1, Tubeimoside A, Tubeimoside I, Tuberimoside 1 |
| InChI Formula | InChI=1S/C63H98O29/c1-26-46-47(88-51-43(76)38(71)30(67)22-81-51)45(78)53(84-26)90-48-39(72)31(68)23-82-54(48)92-56(79)63-15-13-57(2,3)17-28(63)27-9-10-35-59(5)18-29(66)50(60(6,25-65)34(59)11-12-62(35,8)61(27,7)14-16-63)91-55-49(42(75)40(73)32(21-64)86-55)89-52-44(77)41(74)33(24-83-52)85-36(69)19-58(4,80)20-37(70)87-46/h9,26,28-35,38-55,64-68,71-78,80H,10-25H2,1-8H3/t26-,28-,29-,30+,31-,32+,33-,34+,35+,38-,39-,40+,41-,42-,43+,44+,45+,46-,47-,48+,49+,50-,51-,52+,53+,54+,55-,58-,59-,60-,61+,62+,63-/m0/s1 |
| SMILES | C[C@@H]1O[C@@H]2O[C@@H]3[C@@H](O)[C@@H](O)CO[C@@H]3OC(=O)[C@]45CCC(C)(C)C[C@H]4C6=CC[C@@H]7[C@@]8(C)C[C@H](O)[C@H](O[C@@H]9O[C@H](CO)[C@@H](O)[C@H](O)[C@H]9O[C@H]%10OC[C@H](OC(=O)C[C@](C)(O)CC(=O)O[C@@H]1[C@@H](O[C@@H]%11OC[C@@H](O)[C@H](O)[C@H]%11O)[C@H]2O)[C@H](O)[C@H]%10O)[C@@](C)(CO)[C@@H]8CC[C@@]7(C)[C@]6(C)CC5 |
| Molecular Weight (g/mol) | 1319.43 |
| Formula Weight | 1318.6194 |