Learn More
UVP Replacement Tube for UV/UV2 PCR Workstation
Product Code. 12341153
Change view
Click to view available options
Wattage:
15 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage unitSize
12341153 15 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12341153 Supplier UVP Supplier No. 34008701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replace aging light tubes with quality replacement parts. UVP Ultra Violet Product™ Replacement Tube for UV/UV2 PCR Workstation will keep your UV PCR workstation in proper running condition.
  • Suitable for UV/UV2 PCR workstations
  • 15W white light
Especificaciones
Light Source UV
Voltage 230 V
Wattage 15 W
Color White
Electrical Requirements 15 W, 230 V, 50 Hz
For Use With (Equipment) UV/UV2 PCR Workstation
Hertz 50 Hz
Length (Metric) 457 mm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.