Learn More
BD Vacutainer™ Luer Adapters
Product Code. 12967726
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
12967726 Sterile Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12967726 Supplier BD Supplier No. 367300
  • ONLINE EXCLUSIVE PRICE
    260.00 EUR
    254.67 EUR / Pack of 1000
    Save 5.33 EUR
    2% Off
Estimated Shipment: 28-12-2026
Ideal for use with any female luer fitting. BD Vacutainer™ Luer Adapters are sterile devices with a multi-sampling valve that are designed for use with a catheter or hypodermic needle to collect blood using BD Vacutainer™ blood collection tubes.
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides sterile, secure and safe specimen sampling for fast, accurate and consistent results
  • Offers the security of a threaded, locking luer connection, replacing a luer-slip device
  • Latex-free and compatible with a female luer connection or needleless IV site designed for luer lock access
  • Used together with the BD Vacutainer™ single-use tube holder and evacuated blood collection tubes
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Application) Blood collection from a catheter, needleless IV site, hypodermic needle or any female luer fitting; For safe and easy subculturing
For Use With (Equipment) Vacutainer™ Single-use Tube Holders and Vacutainer™ Evacuated Blood Collection Tubes
Includes With luer adapter
Latex Free Yes
Product Type Tube Adapters and Holder