Learn More
Thermo Scientific TUBASSY 8mm x 16mm, NS Y Pump segment 8mm x 16mm Y
Brand:  Thermo Scientific SH3B6531
TUBASSY 8mm x 16mm, NS Y Pump segment 8mm x 16mm Y
Product Code. 17889736
  • ONLINE EXCLUSIVE PRICE
    1155.00 EUR
    950.05 EUR / Pack of 10
    Save 204.95 EUR
    17% Off