missing translation for 'onlineSavingsMsg'
Learn More

TTA-Q6, MedChemExpress

Product Code. 16473908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473908 10 mg 10mg
16490267 50 mg 50mg
16486627 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473908 Supplier Medchem Express Supplier No. HY10388
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TTA-Q6 is a selective T-type Ca2+ channel antagonist, which can be used in the research of neurological disease.

TTA-Q6 is a selective T-type Ca2+ channel antagonist, which can be used in the research of neurological disease.

Specifications

Chemical Name or Material TTA-Q6
Physical Form Solid
CAS 910484-28-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H15ClF3N3O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (308.03 mM; Need ultrasonic)
SMILES N#CC1=CC=C([C@]2(C3CC3)N(CC(F)(F)F)C(NC4=C2C=C(Cl)C=C4)=O)C=C1
Molecular Weight (g/mol) 405.8
Formula Weight 405.8
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.