Learn More
TTA-Q6, MedChemExpress
Product Code. 16490267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490267 50 mg 50mg
16486627 5 mg 5mg
16473908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16490267 Supplier Medchem Express Supplier No. HY10388
MedChemExpress TTA-Q6 is a selective T-type Ca2+ channel antagonist, which can be used in the research of neurological disease.
TTA-Q6 is a selective T-type Ca2+ channel antagonist, which can be used in the research of neurological disease.
Specifications
Chemical Name or Material TTA-Q6
Physical Form Solid
CAS 910484-28-5
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H15ClF3N3O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (308.03 mM; Need ultrasonic)
SMILES N#CC1=CC=C([C@]2(C3CC3)N(CC(F)(F)F)C(NC4=C2C=C(Cl)C=C4)=O)C=C1
Molecular Weight (g/mol) 405.8
Formula Weight 405.8
Percent Purity 99.97%
Grade Research