missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Tryptone Yeast Extract Agar
Description
- ISO
- Solid medium for the enumeration of water microorganisms
- According to ISO 6222, 5667-2, 5667-3, 6887 and 8199 standards.
Specifications
Specifications
| Certifications/Compliance | ISO 6222, 5667-2, 5667-3, 6887 and 8199 |
| Form | Solid |
| Media Type | Tryptone Yeast Extract Agar |
| Product Type | Microbiology Media |
| Formulation | Tryptone 6.00 g/L, Yeast Extract 3.00 g/L, Agar 15.00 g/L |
| For Use With (Application) | Enumeration of Heterotrophic Microorganisms From Water |
| Incubation Conditions | 36°C ± 2.0°C |
| Incubation Time | 44 ± 4 hr. |
| pH | 7.2 ±0.2 |
| Quantity | 500 g |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?