missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- CAS Number: 9002-07-7
- Molecular Weight: Approximately 23,000 or 22000 (for human)
- Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
- Activity: One BAEE unit will produce a DA253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as substrate. Reaction volume = 3.2mL (1 cm light path).
One TAME unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 0.001 M calcium ion.
One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified. - Activity Conversion:1 TAME unit = 19.2 USP or NF units = 57.5 BAEE Units
- Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
Specifications
Specifications
| For Use With (Application) | Release adherent cells from tissue culture plates for passaging |
| Sample Type | Porcine |
| Content And Storage | 4°C |
| Form | Powder |
| Quantity | 25 g |
| Product Type | Trypsin |
Safety and Handling
Signal Word
- Danger
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P321-Specific treatment (see on this label).
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P405-Store locked up.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?