missing translation for 'onlineSavingsMsg'
Learn More

Trypsin inhibitor, MP Biomedicals™

Product Code. 11398699 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
25mg
500mg
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398699 500 mg 500mg
11368699 25 mg 25mg
11378699 100 mg 100mg
11388699 250 mg 250mg
11308709 1 g 1g
11318709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11398699 Supplier MP Biomedicals Supplier No. 0210111390
  • ONLINE EXCLUSIVE PRICE
    595.00 EUR
    565.95 EUR / 500mg
    Save 29.05 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9035-81-8 | Weight: 21500 g/mol

Chemical Identifiers

CAS 9035-81-8
Synonym Ovomucoid trypsin inhibitor

Specifications

Color White
Quantity 500 mg
Physical Form Powder
Chemical Name or Material Trypsin Inhibitor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.