missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ TrueCut™ Cas9 Protein (Prototype)
Description
- Gibco TrueCut Cas9 Protein (Prototype) is our highest performing CRISPR-Cas9 protein, engineered to deliver maximum ribonucleoprotein (RNP)-editing efficiency.
- This protein is provided here in a high-concentration, large-scale format as an early-access material for process development.
- The same high performing enzyme as our TrueCut Cas9 Protein v2, providing superior CRISPR-Cas9 editing efficiency in all tested cell lines, including primary immune cells
- Developed with protocols and demonstrated performance in editing human primary T cells
- Provided at a concentration of 10 mg/mL
- Aseptic manufacturing with sterile filling, bioburden, endotoxin, mycoplasma, residual host nucleic acids, and protein testing
- Designed to meet United States and European Pharmacopoeia standards for Ancillary Materials for Cell, Gene, and Tissue-engineered Products
- TrueCut Cas9 Protein (Prototype) is a Streptococcus pyogenes Cas9 (wt) protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Cas9 Protein |
| Content And Storage | 500 μL TrueCut Cas9 Protein Prototype (10 mg/mL) Store at -15 to -25°C. |
| Shipping Condition | Dry Ice |
| Research Category | Genome Editing |
| Product Line | TrueCut |
For Research Use Only. Not for use in diagnostic procedures. Product is a prototype and performance characteristics of this product have not been established
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?