missing translation for 'onlineSavingsMsg'
Learn More

Tropomodulin 1 Antibody (OTI2C2), Novus Biologicals™

Product Code. 18715673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18715673 Supplier Novus Biologicals Supplier No. NBP200955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tropomodulin 1 Monoclonal specifically detects Tropomodulin 1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomodulin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2C2
Concentration 0.86 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150
Gene Alias D9S57E, Erythrocyte tropomodulin, E-Tmod, e-tropomodulin, TMODETMOD, tropomodulin 1, tropomodulin-1
Gene Symbols TMOD1
Host Species Mouse
Immunogen Full length human recombinant protein of human TMOD1(NP_003266) produced in HEK293T cell.
Molecular Weight of Antigen 40.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7111
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.