Learn More
Multiroir Controlec™ Trolley
Product Code. 15528943
Change view
Click to view available options
Dimensions (L x W x H):
1100 x 520 x 1020 mm (Overall), 504 x 910 mm (Tray)
1100 x 520 x 950 mm (Overall), 504 x 910 mm (Tray)
480 x 800 x 1000 mm (Overall), 435 x 615 mm (Tray)
480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray)
Color:
Black
Light grey
No. of Shelves:
2
3
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Color No. of Shelves Unit Size
15528943 480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray) Light grey 2 Each
15548943 480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray) Black 2 Each
15558943 480 x 800 x 1000 mm (Overall), 435 x 615 mm (Tray) Black 3 Each
15508943 1100 x 520 x 950 mm (Overall), 504 x 910 mm (Tray) Black 2 Each
15598933 1100 x 520 x 1020 mm (Overall), 504 x 910 mm (Tray) Light grey 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15528943 Supplier Multiroir Controlec™ Supplier No. 65005
  • ONLINE EXCLUSIVE PRICE
    191.74 EUR
    177.27 EUR / Pack of 1
    Save 14.47 EUR
    7% Off
Estimated Shipment: 20-10-2026
The Multiroir multi-purpose plastic trolleys are practicle and functional, ideal for general use, transporting and moving multiple items around the lab or workspace.
  • Aluminum frame with handles at each end
  • Available in two or three tray format
  • On casters for easy manouvrability
  • Available in light grey or black
Specifications
Color Light grey
Dimensions (L x W x H) 480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray)
Load Bearing Capacity (Metric) 180 kg (Overall)
Material Aluminum Frame, Polypropylene Trays
No. of Shelves 2
Shelf Material Polypropylene
Wheel Type Lockable casters