Learn More
Trisodium Naphthalene-1,3,6-trisulfonate, 65%, BLD Pharm™
Specifications
Specifications
| CAS | 5182-30-9 |
| Molecular Formula | C10H5Na3O9S3 |
| MDL Number | MFCD00003989 |
| Quantity | 100 g |
| SMILES | O=S(C1=C2C=CC(S(=O)([O-])=O)=CC2=CC(S(=O)([O-])=O)=C1)([O-])=O.[Na+].[Na+].[Na+] |
| Molecular Weight (g/mol) | 434.31 |
| Percent Purity | 65% |
| Chemical Name or Material | Trisodium Naphthalene-1,3,6-trisulfonate |