Learn More
Tris(trimethylsilyl)guanine, TRC
Product Code. 30455495
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455495 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455495 Supplier Toronto Research Chemicals Supplier No. TRCT88651010G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris(trimethylsilyl)guanine
CAS 18602-85-2
Molecular Formula C14H29N5OSi3
Quantity 10 g
Synonym N,9-Bis(trimethylsilyl)-6-[(trimethylsilyl)oxy]-9H-purin-2-amine, 2,6,9-Tris(trimethylsilyl)guanine, 6-(Trimethylsiloxy)-9-(trimethylsilyl)-2-[(trimethylsilyl)amino]-9H-purine
InChI Formula InChI=1S/C14H29N5OSi3/c1-21(2,3)18-14-16-12-11(13(17-14)20-23(7,8)9)15-10-19(12)22(4,5)6/h10H,1-9H3,(H,16,17,18)
SMILES C[Si](C)(C)Nc1nc(O[Si](C)(C)C)c2ncn(c2n1)[Si](C)(C)C
IUPAC Name N,9-bis(trimethylsilyl)-6-trimethylsilyloxypurin-2-amine
Molecular Weight (g/mol) 367.67
Formula Weight 367.168