missing translation for 'onlineSavingsMsg'
Learn More

Tris(diethylamino)gallium(III), 99.9% (metals basis), Thermo Scientific™

Product Code. 11335567
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335567 500 mg 500mg
11325567 2 g 2g
11315567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335567 Supplier Thermo Scientific Alfa Aesar Supplier No. 042759.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194611-64-8
Molecular Formula C12H30GaN3
Molecular Weight (g/mol) 286.116
MDL Number MFCD01863475
InChI Key UZYORTJMAUFROH-UHFFFAOYSA-N
Synonym tris diethylamino gallium iii
PubChem CID 134068145
IUPAC Name gallium;diethylazanide
SMILES CC[N-]CC.CC[N-]CC.CC[N-]CC.[Ga+3]

Specifications

Color Brown
Linear Formula Ga[N(CH2CH3)2]3
Quantity 500 mg
UN Number UN3263
Sensitivity Moisture sensitive
Formula Weight 286.11
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tris (diethylamino)gallium (III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.