Learn More
Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)
Product Code. 11332249
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332249 5 g 5g
11322249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11332249 Supplier Thermo Scientific Alfa Aesar Supplier No. H27238.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1272-25-9 | C15H15Tb | 354.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1272-25-9
Molecular Formula C15H15Tb
Molecular Weight (g/mol) 354.21
MDL Number MFCD03427139
InChI Key WFRJBVQIASPUDD-UHFFFAOYSA-N
Synonym tris cyclopentadienyl terbium, acmc-1bvr0, tris, c5-cyclopenta-2,4-dien-1-yl terbium
PubChem CID 20025873
IUPAC Name cyclopenta-1,3-diene;terbium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Tb+3]
Specifications
Melting Point 235°C
Quantity 5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 354.22
Percent Purity 99%
Assay (99.9%-Tb)
Chemical Name or Material Tris(cyclopentadienyl)terbium(III), (REO)