accessibility menu, dialog, popup

UserName

CAS RN 1272-25-9

Tb

CAS RN 1272-25-9

IUPAC Name: tris(cyclopenta-2,4-dien-1-yl)terbium
Synonyms: tris(cyclopenta-2,4-dien-1-yl)terbium
Molecular Weight (g/mol): 354.21
Molecular Formula: C15H15Tb
InChi Key: FMNCAPFXHVPBJH-UHFFFAOYSA-N
SMILES: C1=CC(C=C1)[Tb](C1C=CC=C1)C1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results