Learn More
TrippNT Bottle Holders
Product Code. 11776758 Shop All Cole Parmer Products
Change view
Click to view available options
Holds:
Squeeze Bottles (500 mL)
Positions:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Positions Unit Size
11776758 Squeeze Bottles (500 mL) 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11776758 Supplier TrippNT Supplier No. 6352640
Estimated Shipment: 22-12-2026
Avoid messy spills or accidents with these bottle holders. Wash Bottle Holders keep wash solutions organized and close at hand.
Avoid messy spills or accidents with these bottle holders. Wash Bottle Holders keep wash solutions organized and close at hand.
  • Bottle holders for 500mL or 1L squeeze bottles
  • Mount to wall, hood, or benchtop using attached double-faced tape
  • Sturdy, PVC construction
  • Accommodates standard wash bottles
Specifications
Holds Squeeze Bottles (500 mL)
Material Chemical-resistant Plastic
Positions 1
Product Type Bottle Holder