Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 100μL
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
100 μL
missing translation for 'unitSize'
Each
Description
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
Specifications
| Needle Gauge | 23s G |
| Product Type | Fixed Gastight Syringe |
| Volume (Metric) | 100 μL |
| Needle Point Style | Cone |