missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine, Sigma-Aldrich™

Product Code. 15884241
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884241 1 kg 1kg
15864241 5 g 5g
15874241 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15884241 Supplier Sigma-Aldrich Supplier No. 8082701000
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-35-0

Specifications

Melting Point 78.5°C to 81.5°C
Density 1.194 g/cm3 at 20°C
Boiling Point 377°C/1013 hPa
Molecular Formula (C6H5)3P
MDL Number MFCD00003043
Quantity 1 kg
Synonym Triphenylphosphine, Phosphorus triphenyl, Triphenylphosphine
Molecular Weight (g/mol) 262.29
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.