Learn More
Triphenylphosphine Oxide-d15, TRC
Especificaciones
Especificaciones
| Chemical Name or Material | Triphenylphosphine Oxide-d15 |
| CAS | 54964-93-1 |
| Molecular Formula | C18D15OP |
| Quantity | 50 mg |
| Synonym | Tri(phenyl-d5)phosphine Oxide, NSC 398-d15, PP 560-d15, TPPO-d15, Triphenyl Phosphorus Oxide-d15, Triphenylphosphine Monoxide-d15, Orlistat Related Compound C-d15, Orlistat USP Related Compound C-d15 |
| InChI Formula | InChI=1S/C18H15OP/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D |
| SMILES | P(=O)(C1=C(C(=C(C(=C1[2H])[2H])[2H])[2H])[2H])(C2=C(C(=C(C(=C2[2H])[2H])[2H])[2H])[2H])C3=C(C(=C(C(=C3[2H])[2H])[2H])[2H])[2H] |
| IUPAC Name | tris(phenyl-d5)phosphine oxide |
| Molecular Weight (g/mol) | 293.38 |
| Formula Weight | 293.18 |