Learn More
Triphenyl Phosphate, Sigma-Aldrich™
Product Code. 15891441
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891441 100 g 100g
15801451 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15891441 Supplier Sigma Aldrich Supplier No. 8211970100
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 115-86-6
Specifications
Melting Point 48°C to 50°C
Boiling Point 220°C/5 hPa
Molecular Formula (C6H5O)3PO
MDL Number MFCD00003031
Quantity 100 g
Synonym Triphenyl phosphate, Phosphoric acid triphenyl ester, Triphenyl phosphate
Molecular Weight (g/mol) 326.28
Grade Research