missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin, ∽99%, MP Biomedicals™

Product Code. 11487621 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487621 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487621 Supplier MP Biomedicals Supplier No. 0210479425

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 555-45-3
Color White
Assay Percent Range ∽99%
Molecular Formula C45H86O6
Quantity 25 g
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
Molecular Weight (g/mol) 723.177
PubChem CID 11148
ChEBI CHEBI:77391
Physical Form Crystalline Powder
Chemical Name or Material Trimyristin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.