Learn More
Trimethylsulfonium Hydroxide (0.2mol/L in Methanol), TRC
Product Code. 30455850
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455850 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455850 Supplier Toronto Research Chemicals Supplier No. TRCT7970001ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimethylsulfonium Hydroxide
CAS 17287-03-5
Molecular Formula C3H10OS
Quantity 1 mL
Synonym Sulfonium','trimethyl-','hydroxide (8CI','9CI), Trimethylsulfonium hydroxide (6CI)
InChI Formula InChI=1S/C3H9S.H2O/c1-4(2)3;/h1-3H3;1H2/q+1;/p-1
SMILES [OH-].C[S+](C)C
IUPAC Name trimethylsulfanium;hydroxide
Molecular Weight (g/mol) 94.18
Formula Weight 94.0452
Concentration or Composition (by Analyte or Components) 0.2 mol/L