Learn More
Trimethylphenyltin, TRC
Specifications
Specifications
| Chemical Name or Material | Trimethylphenyltin |
| CAS | 934-56-5 |
| Molecular Formula | C9H14Sn |
| Quantity | 10 g |
| Synonym | Phenyltrimethylstannane, Phenyltrimethyltin, Trimethylmonophenyltin, Trimethylphenylstannane, Trimethylstannylbenzene |
| InChI Formula | InChI=1S/C6H5.3CH3.Sn/c1-2-4-6-5-3-1;;;;/h1-5H;3*1H3; |
| SMILES | C[Sn](C)(C)c1ccccc1 |
| IUPAC Name | trimethyl(phenyl)stannane |
| Molecular Weight (g/mol) | 240.92 |
| Formula Weight | 242.0117 |