missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl-d9-acetic Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Pivalic acid-d9 |
| CAS | 42983-07-3 |
| Molecular Formula | C5 D9 H O2 |
| Quantity | 0.5 g |
| Synonym | 2,2-Di(methyl-d3)propanoic-3,3,3-d3 Acid, 2,2-Dimethylpropanoic Acid-d9, 2,2,2-Trimethylacetic Acid-d9, 2,2-Dimethylpropanoic Acid-d9, 2,2-Dimethylpropionic Acid-d9, NSC 65449-d9, Neopentanoic Acid-d9, Neovaleric Acid-d9, Trimethylacetic Acid-d9, Trimethylmethanecarboxylic Acid-d9, Versatic 5 Acid-d9, tert-Pentanoic Acid-d9, alpha,alpha-Dimethylpropionic Acid-d9, Trimethyl-d9-acetic Acid |
| InChI Formula | InChI=1 S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,3-trideuterio-2,2-bis(trideuteriomethyl)propanoic acid |
| Molecular Weight (g/mol) | 111.19 |
| Formula Weight | 111.1246 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?