missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethyl 1,2,4-benzenetricarboxylate, 98%

Product Code. 11415917
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415917 500 g 500g
11495907 25 g 25g
11405917 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415917 Supplier Alfa Aesar™ Supplier No. B24455.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10-1-2459
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00043629
InChI Key MJHNUUNSCNRGJE-UHFFFAOYSA-N

Specifications

Melting Point 38°C to 40°C
Boiling Point 193°C to 195°C (12 mmHg)
Flash Point 182°C (360°F)
Quantity 500 g
Synonym trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)C(=O)OC
IUPAC Name trimethyl benzene-1,2,4-tricarboxylate
PubChem CID 17159
Formula Weight 252.22
Percent Purity 98%
Chemical Name or Material Trimethyl 1,2,4-benzenetricarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.