missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | TRIM6 |
|---|---|
| Concentration | 0.22 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859363
|
Proteintech
11953-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849363
|
Proteintech
11953-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| TRIM6 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q8BGE7, Q9C030 | |
| 117854, 293294, 94088 | |
| TRIM6 Fusion Protein Ag2556 | |
| Primary | |
| -20°C | |
| TRIM6 |
| 0.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| RING finger protein 89, RNF89, TRIM6, tripartite motif containing 6 | |
| Trim6 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title