missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11953-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| TRIM6 | |
| Polyclonal | |
| Unconjugated | |
| TRIM6 | |
| RING finger protein 89, RNF89, TRIM6, tripartite motif containing 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117854, 293294, 94088 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BGE7, Q9C030 | |
| Trim6 | |
| TRIM6 Fusion Protein Ag2556 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction