missing translation for 'onlineSavingsMsg'
Learn More

Trihydroxycoprostane 3,7,12-Triethoxymethyl Ether, TRC

Product Code. 30456543
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456543 50 mg 50mg
30452479 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456543 Supplier Toronto Research Chemicals Supplier No. TRCT79516050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trihydroxycoprostane 3,7,12-Triethoxymethyl Ether
Analyte or Component Names Trihydroxycoprostane 3,7,12-Triethoxymethyl Ether
Shipping Condition Room Temperature
Molecular Formula C36 H66 O6
InChI Formula InChI=1S/C36H66O6/c1-9-37-22-40-28-17-18-35(7)27(19-28)20-32(41-23-38-10-2)34-30-16-15-29(26(6)14-12-13-25(4)5)36(30,8)33(21-31(34)35)42-24-39-11-3/h25-34H,9-24H2,1-8H3/t26-,27+,28-,29-,30+,31+,32-,33+,34+,35+,36-/m1/s1
Solution Type Neat
SMILES CCOCO[C@@H]1CC[C@@]2(C)[C@@H](C1)C[C@@H](OCOCC)[C@H]3[C@@H]4CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)[C@H](C[C@H]23)OCOCC
IUPAC Name (3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-tris(ethoxymethoxy)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
Industry Type Pharmaceutical
Molecular Weight (g/mol) 594.906
Quantity 50 mg
Formula Weight 594.486
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.