Learn More
Triglycidyl Isocyanurate-d15, TRC
Specifications
Specifications
| Chemical Name or Material | Triglycidyl Isocyanurate-d15 |
| Molecular Formula | C12D15N3O6 |
| Quantity | 5 mg |
| Synonym | 1,3,5-Triglycidyl isocyanurate-d15, 1,3,5-Triglycidyl-s-triazine-2,4,6-trione-d15, 1,3,5-Triglycidylhexahydro-1,3,5-triazine-2,4,6-trione-d15, 1,3,5-Triglycidylisocyanuric acid-d15, 1,3,5-Tris(2,3-epoxypropyl) isocyanurate-d15, 1,3,5- Tris(oxiran-2-ylmethyl)-1,3,5-triazine-2,4,6-trione-d15, 1,3,5-Tris(oxiranylmethyl)-1,3,5-triazine-2,4,6-trione-d15, Glycidyl Isocyanurate-d15, N,N',N''-Triglycidyl isocyanurate-d15, NSC 269934, PTGIC-d15, TGT-d15, Triglycidyl isocyanurate-d15, Tris(2,3-epoxypropyl) Isocyanurate-d15, Tris(epoxypropyl) Isocyanurate-d15 |
| InChI Formula | InChI=1S/C12H15N3O6/c16-10-13(1-7-4-19-7)11(17)15(3-9-6-21-9)12(18)14(10)2-8-5-20-8/h7-9H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D |
| SMILES | O=C(N(C([2H])([2H])C1([2H])C([2H])([2H])O1)C(N2C([2H])([2H])C3([2H])C([2H])([2H])O3)=O)N(C([2H])([2H])C4([2H])C([2H])([2H])O4)C2=O |
| Molecular Weight (g/mol) | 312.36 |