missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic Acid, MP Biomedicals

Product Code. 11449481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449481 10 g 10g
11459481 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449481 Supplier MP Biomedicals Supplier No. 0215702510
  • ONLINE EXCLUSIVE PRICE
    52.95 EUR
    50.40 EUR / 10g
    Save 2.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Free Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
IUPAC Name trifluoromethanesulfonic acid
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Quantity 10 g
Formula Weight 150.1
Concentration or Composition (by Analyte or Components) 1mL = approx. 1.70g
Chemical Name or Material Trifluoromethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.