missing translation for 'onlineSavingsMsg'
Learn More

Trifloxystrobin, TRC

Product Code. 17932005
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932005 250 mg 250mg
30133222 100 mg 100mg
17960425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932005 Supplier Toronto Research Chemicals Supplier No. TRCT778900250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141517-21-7
Molecular Formula C20 H19 F3 N2 O4
Molecular Weight (g/mol) 408.37
Synonym Benzeneacetic acid, α-(methoxyimino)-2-[[[(E)-[1-[3-(trifluoromethyl)phenyl]ethylidene]amino]oxy]methyl]-, methyl ester, (αE)-, Benzeneacetic acid, α-(methoxyimino)-2-[[[[1-[3-(trifluoromethyl)phenyl]ethylidene]amino]oxy]methyl]-, methyl ester, (E,E)-, CGA 279202, Compass, Compass (strobilurin fungicide), Consist, Flint, Flint (fungicide), Tega, Trifloxystrobin, Trilex, Twist, Zato, Zato 50WG
IUPAC Name methyl (2E)-2-methoxyimino-2-[2-[[(E)-1-[3-(trifluoromethyl)phenyl]ethylideneamino]oxymethyl]phenyl]acetate
SMILES CO\N=C(\C(=O)OC)/c1ccccc1CO\N=C(/C)\c2cccc(c2)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H19F3N2O4/c1-13(14-8-6-9-16(11-14)20(21,22)23)24-29-12-15-7-4-5-10-17(15)18(25-28-3)19(26)27-2/h4-11H,12H2,1-3H3/b24-13+,25-18+
Formula Weight 408.1297
Chemical Name or Material Trifloxystrobin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.