missing translation for 'onlineSavingsMsg'
Learn More

Trifloxystrobin, MedChemExpress

Product Code. 16495816
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495816 100 mg 100mg
16471327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495816 Supplier Medchem Express Supplier No. HY123230
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Trifloxystrobin (CGA 279202) is a fungicide, with EC50s of 23.0 μg/L and 1.7 μg/L for Daphnia magna neonate and embryos, respectively, after treatment for 48 h.

Trifloxystrobin (CGA 279202) is a fungicide, with EC50s of 23.0 μg/L and 1.7 μg/L for Daphnia magna neonate and embryos, respectively, after treatment for 48 h.

Specifications

Chemical Name or Material Trifloxystrobin
Physical Form Solid
CAS 141517-21-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H19F3N2O4
For Use With (Application) COVID-19-immunoregulation
Synonym CGA 279202
Solubility Information DMSO : 125 mg/mL (306.09 mM; Need ultrasonic)
SMILES C/C(C1=CC(C(F)(F)F)=CC=C1)=N\OCC2=C(C=CC=C2)/C(C(OC)=O)=N\OC
Molecular Weight (g/mol) 408.37
Formula Weight 408.37
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.