Learn More
Trifloxystrobin Acid, TRC
Product Code. 30007242
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007242 50 mg 50mg
30009056 10 mg 10mg
30004797 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007242 Supplier Toronto Research Chemicals Supplier No. TRCT13818550MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 252913-85-2
Molecular Formula C19 H17 F3 N2 O4
Molecular Weight (g/mol) 394.34
Synonym (αE)-α-(Methoxyimino)-2-[[[(E)-[1-[3-(trifluoromethyl)phenyl]ethylidene]amino]oxy]methyl]benzeneacetic acid, (E,E)-α-(Methoxyimino)-2-[([1-[3-(trifluoromethyl)phenyl]ethylidene]amino)oxymethyl]benzeneacetic acid, CGA 321113, Trifloxystrobin acid
IUPAC Name (2E)-2-methoxyimino-2-[2-[[(E)-1-[3-(trifluoromethyl)phenyl]ethylideneamino]oxymethyl]phenyl]acetic acid
SMILES CO\N=C(\C(=O)O)/c1ccccc1CO\N=C(/C)\c2cccc(c2)C(F)(F)F
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C19H17F3N2O4/c1-12(13-7-5-8-15(10-13)19(20,21)22)23-28-11-14-6-3-4-9-16(14)17(18(25)26)24-27-2/h3-10H,11H2,1-2H3,(H,25,26)/b23-12+,24-17+
Formula Weight 394.114
Percent Purity >95%
Chemical Name or Material Trifloxystrobin (free acid)
Grade HPLC